missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLTF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582525
This item is not returnable.
View return policy
Description
Immunogen sequence: LKKHGFKLGP APKTLGFNLE SGWGSGRAGP SYSMPVHAAV QMTTEQLKTE FDKLFEDLKE DDKTHEMEPA EAIETPLLPH QKQALAWMVS RENSKELPPF WEQRNDLYYN TITNFSEKDR PENVHGGILA DDMGLGK Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 89%.
This gene encodes a member of the SWI/SNF family. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein contains a RING finger DNA binding motif. Two transcript variants encoding the same protein have been found for this gene. However, use of an alternative translation start site produces an isoform that is truncated at the N-terminus compared to the full-length protein.
Specifications
| HLTF | |
| Polyclonal | |
| Unconjugated | |
| HLTF | |
| AF010600; BC057116; DNA-binding protein/plasminogen activator inhibitor 1 regulator; DNA-binding protein/plasminogen activator inhibitor-1 regulator; helicase like transcription factor; helicase-like transcription factor; HIP116; HIP116A; HLTF; HLTF1; mem; P113; RING finger protein 80; RING-type E3 ubiquitin transferase HLTF; RNF80; Smarca3; Snf2l3; SNF2-like 3; Sucrose nonfermenting protein 2-like 3; sucrose nonfermenting-like 3; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 3; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 3; TNF-response element-binding protein; ZBU1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20585, 295568, 6596 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14527, Q6PCN7 | |
| HLTF | |
| Recombinant protein corresponding to Human HLTF. Recombinant protein control fragment (Product #RP-91601). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction