missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £309.00
Specifications
| Antigen | HLM |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18125472
|
Novus Biologicals
NBP2-23625SS |
0.025 mg |
£155.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18106601
|
Novus Biologicals
NBP2-23625 |
0.1 mg |
£309.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLM Polyclonal specifically detects HLM in Human samples. It is validated for Western Blot.Specifications
| HLM | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 23762 | |
| IgG | |
| Affinity Purified |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| ORP4, ORP-4, OSBPL1, OSBPL4 | |
| Reacts with residues 549-563 [EKGRWDEANTTEKQR] of the HLM protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title