missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12870-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the proline and acidic-rich (PAR) protein family, a subset of the bZIP transcription factors. The encoded protein forms homodimers or heterodimers with other PAR family members and binds sequence-specific promoter elements to activate transcription. Chromosomal translocations fusing portions of this gene with the E2A gene cause a subset of childhood B-lineage acute lymphoid leukemias. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Specifications
| HLF | |
| Polyclonal | |
| Unconjugated | |
| HLF | |
| hepatic leukemia factor, HLF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 217082, 3131, 690286 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16534, Q64709, Q8BW74 | |
| HLF | |
| HLF Fusion Protein Ag3533 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction