missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102942
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HLF.
This gene encodes a member of the proline and acidic-rich (PAR) protein family, a subset of the bZIP transcription factors. The encoded protein forms homodimers or heterodimers with other PAR family members and binds sequence-specific promoter elements to activate transcription. Chromosomal translocations fusing portions of this gene with the E2A gene cause a subset of childhood B-lineage acute lymphoid leukemias. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
| HLF | |
| Polyclonal | |
| Unconjugated | |
| HLF | |
| E230015K02Rik; hepatic leukemia factor; HLF; HLF transcription factor, PAR bZIP family member; HLF, PAR bZIP transcription factor; LOC690286; PAR bZIP transcription factor; similar to hepatic leukemia factor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 217082, 3131 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16534, Q8BW74 | |
| HLF | |
| A synthesized peptide derived from human HLF(Accession Q16534), corresponding to amino acid residues T73-V123. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction