missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-G Monoclonal Antibody (MEM-G/9), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA119643
This item is not returnable.
View return policy
Description
This antibody reacts with an extracellular epitope on native form of human HLA-G1 on the cell surface as well as with soluble HLA-G5 isoform in its beta2-microglobulin associated form. Reactivity with HLA-G3 was also reported. It will not cross-react with mouse.
HLA-G belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-G is expressed on fetal derived placental cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exon 6 encodes the cytoplasmic tail. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| HLA-G | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
| P17693 | |
| HLA-G | |
| Recombinant human HLA-G refolded with beta- 2 Microglobulin and peptide. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| MEM-G/9 | |
| PE | |
| HLA-G | |
| acrosomal protein SP-10; Acrosomal vesicle protein 1; ACRV1; b2 microglobulin; D11S4365; DADB-15K14.8; HLA class I histocompatibility antigen, alpha chain G; HLA class I molecule; HLA G antigen; HLA-6.0; HLAG; HLA-G; HLA-G histocompatibility antigen, class I, G; LOC768350; major histocompatibility complex, class I, G; MHC class I antigen G; MHC-G; MHCIY; MHC-like class I Y; sHLA-G; Soluble HLA class I histocompatibility antigen, alpha chain G; SP-10; SPACA2; sperm protein 10 | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| 3135 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction