missing translation for 'onlineSavingsMsg'
Learn More

HLA G Antibody (2A12), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18423223 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423223 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423223 Supplier Novus Biologicals Supplier No. NBP144925MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HLA G Monoclonal antibody specifically detects HLA G in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA G
Applications Flow Cytometry
Classification Monoclonal
Clone 2A12
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC class Ib antigen, MHC-G, mutant MHC class I antigen, mutant MHC class Ib antigen, sHLA-G, Soluble HLA class I histocompatibility antigen, alpha chain G
Host Species Mouse
Immunogen C-terminal amino acid sequence (22-mer) of soluble HLA-G5 and HLA-G6 proteins coupled to ovalbumin (NM_002127.5).
Purification Method Multi-step
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 3135
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.