missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HLA-F Recombinant Rabbit Monoclonal Antibody (24GB3830)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557038

 View more versions of this product

Product Code. 30275647

  • £126.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HLA-F Recombinant Monoclonal Antibody for Western Blot, ICC/IF

HLA-F belongs to the HLA class I heavy chain paralogues. It is a non-classical heavy chain that forms a heterodimer with a beta-2 microglobulin light chain, with the heavy chain anchored in the membrane. Unlike most other HLA heavy chains, this molecule is localized in the endoplasmic reticulum and Golgi apparatus, with a small amount present at the cell surface in some cell types. It contains a divergent peptide-binding groove, and is thought to bind a restricted subset of peptides for immune presentation. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HLA-F
Recombinant Monoclonal
0.387 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P30511
HLA-F
A synthesized peptide derived from human HLA F (aa 25-76).
20 μL
Primary
Human
Antibody
IgG
Western Blot, Immunocytochemistry
24GB3830
Unconjugated
HLA-F
CDA12; HLA class I histocompatibility antigen, alpha chain F; HLA class I molecule; HLA F antigen; HLA-5.4; HLA-CDA12; HLAF; HLA-F; Leukocyte antigen F; major histocompatibility complex, class I, F; MHC class I antigen F; MHC class Ib antigen
Rabbit
Affinity chromatography
RUO
3134
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.