missing translation for 'onlineSavingsMsg'
Learn More

HLA-F Mouse anti-Human, Clone: 1B12C2, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen HLA-F
Clone 1B12C2
Concentration 0.94 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16869045
View Documents
Proteintech
66819-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16859045
View Documents
Proteintech
66819-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HLA-F belongs to the HLA class I heavy chain paralogues. It is a non-classical heavy chain that forms a heterodimer with a beta-2 microglobulin light chain, with the heavy chain anchored in the membrane. Unlike most other HLA heavy chains, this molecule is localized in the endoplasmic reticulum and Golgi apparatus, with a small amount present at the cell surface in some cell types. It contains a divergent peptide-binding groove, and is thought to bind a restricted subset of peptides for immune presentation.
Specifications

Specifications

HLA-F
0.94 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CDA12, HLA 5.4, HLA CDA12, HLA F, HLA F antigen, HLAF, HLA-F, Leukocyte antigen F, MHC class I antigen F
HLA-F
IgG1
Protein G
Antibody
1B12C2
Western Blot
Unconjugated
Mouse
Human
P30511
3134
HLA-F Fusion Protein Ag6475
Primary
-20°C
HLA-F
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.