missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-F Monoclonal Antibody (3F11F4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548491
This item is not returnable.
View return policy
Description
HLA-F Monoclonal Antibody for ELISA, Flow, Western Blot
HLA-F belongs to the HLA class I heavy chain paralogues. It is a non-classical heavy chain that forms a heterodimer with a beta-2 microglobulin light chain, with the heavy chain anchored in the membrane. Unlike most other HLA heavy chains, this molecule is localized in the endoplasmic reticulum and Golgi apparatus, with a small amount present at the cell surface in some cell types. It contains a divergent peptide-binding groove, and is thought to bind a restricted subset of peptides for immune presentation. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| HLA-F | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P30511 | |
| HLA-F | |
| Purified recombinant fragment of human HLA-F (AA: 22-305) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Western Blot | |
| 3F11F4 | |
| Unconjugated | |
| HLA-F | |
| CDA12; HLA class I histocompatibility antigen, alpha chain F; HLA class I molecule; HLA F antigen; HLA-5.4; HLA-CDA12; HLAF; HLA-F; Leukocyte antigen F; major histocompatibility complex, class I, F; MHC class I antigen F; MHC class Ib antigen | |
| Mouse | |
| Protein G | |
| RUO | |
| 3134 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction