missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA F Antibody (SR1707), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22318-100ul
This item is not returnable.
View return policy
Description
HLA F Monoclonal antibody specifically detects HLA F in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| HLA F | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1707 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| alpha chain F, HLA class I molecule, HLA F antigen, HLA-5.4, HLAFHLA-CDA12, Leukocyte antigen F, major histocompatibility complex, class I, F, MHC class I antigen F, MHC class Ib antigen | |
| A synthesized peptide derived from human HLA F (Uniprot #: P30511) | |
| 100 μg | |
| Immunology | |
| 3134 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction