missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA F Antibody (3D11), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76802AF532
This item is not returnable.
View return policy
Description
HLA F Monoclonal specifically detects HLA F in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| HLA F | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HLA-F | |
| Inclusion body-derived HLA-F heavy chain | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| 3D11 | |
| Western Blot, Flow Cytometry | |
| alpha chain F, HLA class I molecule, HLA F antigen, HLA-5.4, HLAFHLA-CDA12, Leukocyte antigen F, major histocompatibility complex, class I, F, MHC class I antigen F, MHC class Ib antigen | |
| Mouse | |
| 0.1 mL | |
| 3134 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction