missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-E Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15008-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody MEM-E/08 recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. The published results
The antibody MEM-E/08 recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. The published results revealed that antibody is remarkably specific for HLA-E, only with weak cross-reactivity with following classical MHC Class I molecules (MHC Class Ia): HLA-A24, -B7, -B27, -B51, -B54, -C7.Specifications
| HLA-E | |
| Polyclonal | |
| Unconjugated | |
| HLA-E | |
| EA1.2, EA2.1, HLA 6.2, HLA E, HLAE, HLA-E, MHC, MHC class I antigen E, QA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3133 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13747 | |
| HLA-E | |
| HLA-E Fusion Protein Ag6703 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction