missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-E Antibody (MEM-E/07), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-310MFV500
This item is not returnable.
View return policy
Description
HLA-E Monoclonal antibody specifically detects HLA-E in Human samples. It is validated for Flow Cytometry
Specifications
| HLA-E | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| MEM-E/07 | |
| Flow Cytometry | |
| DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
| Bacterially expressed recombinant HLA-E refolded with beta2-microglobulin and peptide. | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Diabetes Research, Immunology | |
| 3133 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction