missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-E Antibody (MEM-E/07), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-310G
This item is not returnable.
View return policy
Description
HLA-E Monoclonal specifically detects HLA-E in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HLA-E | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HLA-E | |
| Bacterially expressed recombinant HLA-E refolded with beta2-microglobulin and peptide. | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
| 3133 | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MEM-E/07 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody MEM-E/07 recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. The published results revealed that the antibody cross-reacts with some classical MHC Class I molecules (MHC Class Ia): HLA-B7 (strongly), HLA-B8 (moderately), HLA-B27, -B44 (weakly). | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction