missing translation for 'onlineSavingsMsg'
Learn More

HLA-E Antibody (HLAE/9468), Novus Biologicals™

Mouse Monoclonal Antibody

£195.00 - £368.00

Specifications

Antigen HLA-E
Clone HLAE/9468
Concentration 0.2 mg/ml
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30290746
View Documents
Novus Biologicals
NBP3-44604-20ug
20 μg
£195.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30289428
View Documents Promotion Details
Novus Biologicals
NBP3-44604-100ug
100 μg
£368.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HLA-E Monoclonal antibody specifically detects HLA-E in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

HLA-E
0.2 mg/ml
Monoclonal
Purified
RUO
Human
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Recombinant fragment (around aa1-150) of human HLA-E protein (Uniprot: P13747). Exact sequence is proprietary.
Primary
Store at 4⪚C.
HLAE/9468
Immunohistochemistry
Unconjugated
Mouse
Adaptive Immunity, Asthma, Diabetes Research, Immunology
10 mM PBS with 0.05% BSA
3133
IgG2 κ
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado