missing translation for 'onlineSavingsMsg'
Learn More

HLA-E Antibody (1079755) [DyLight 405], Novus Biologicals™

Product Code. 30763711 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763711 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763711 Supplier Novus Biologicals Supplier No. FAB11547E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HLA-E Monoclonal antibody specifically detects HLA-E in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-E
Applications Immunohistochemistry
Classification Monoclonal
Clone 1079755
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Host Species Mouse
Immunogen HLA-E containing synthetic peptide, Accession # P13747
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3133
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.