missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DRB5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | HLA DRB5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18316196
|
Novus Biologicals
NBP3-03663-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18377466
|
Novus Biologicals
NBP3-03663-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA DRB5 Polyclonal specifically detects HLA DRB5 in Human samples. It is validated for Western Blot.Specifications
| HLA DRB5 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DR beta-5, DR2-beta-2, DR9, DR-9, dw2, FLJ76359, HLA class II histocompatibility antigen, DR beta 5 chain, HLA class II histocompatibility antigen, DR-5 beta chain, HLA class II histocompatibility antigen, DRB1-9 beta chain, HLA-DRB, human leucocyte antigen DRB5, leukocyte antigen class II, major histocompatibility complex, class II, DR beta 5, MHC class II antigen DRB1*9, MHC class II antigen DRB5, MHC class II HLA beta chain, MHC HLA-DR-beta cell surface glycoprotein, MHC HLA-DR-beta chain | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-227 of human HLA DRB5 (NP_002116.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Asthma, Diabetes Research, Immunology | |
| PBS with 50% glycerol, pH7.3. | |
| 3127 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title