missing translation for 'onlineSavingsMsg'
Learn More

HLA DRB5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18365996 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365996 100 μL 100µL
18343185 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18365996 Supplier Novus Biologicals Supplier No. NBP303664100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HLA DRB5 Polyclonal specifically detects HLA DRB5 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA DRB5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias DR beta-5, DR2-beta-2, DR9, DR-9, dw2, FLJ76359, HLA class II histocompatibility antigen, DR beta 5 chain, HLA class II histocompatibility antigen, DR-5 beta chain, HLA class II histocompatibility antigen, DRB1-9 beta chain, HLA-DRB, human leucocyte antigen DRB5, leukocyte antigen class II, major histocompatibility complex, class II, DR beta 5, MHC class II antigen DRB1*9, MHC class II antigen DRB5, MHC class II HLA beta chain, MHC HLA-DR-beta cell surface glycoprotein, MHC HLA-DR-beta chain
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-227 of human HLA DRB5 (NP_002116.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Asthma, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3127
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.