missing translation for 'onlineSavingsMsg'
Learn More

HLA DRB5 Antibody, Novus Biologicals™

Product Code. 18345534 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345534 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345534 Supplier Novus Biologicals Supplier No. NBP313003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HLA DRB5 Polyclonal antibody specifically detects HLA DRB5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA DRB5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias DR beta-5, DR2-beta-2, DR9, DR-9, dw2, FLJ76359, HLA class II histocompatibility antigen, DR beta 5 chain, HLA class II histocompatibility antigen, DR-5 beta chain, HLA class II histocompatibility antigen, DRB1-9 beta chain, HLA-DRB, human leucocyte antigen DRB5, leukocyte antigen class II, major histocompatibility complex, class II, DR beta 5, MHC class II antigen DRB1*9, MHC class II antigen DRB5, MHC class II HLA beta chain, MHC HLA-DR-beta cell surface glycoprotein, MHC HLA-DR-beta chain
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human HLA DRB5. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Asthma, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3127
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.