missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-DRB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102439
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HLA-DRB3.
Major histocompatibility complex (MHC) class II molecules destined for presentation to CD4+ helper T cells is determined by two key events. These events include the dissociation of class II-associated invariant chain peptides (CLIP) from an antigen binding groove in mhc ii-a/b dimers through the activity of MHC molecules HLA-DM and -DO, and subsequent peptide antigen binding. Accumulating in endosomal/lysosomal compartments and on the surface of B cells, HLA-DM, -DO molecules regulate the dissociation of CLIP and the subsequent binding of exogenous peptides to HLA class II molecules (HLA-DR, -DQ and -DP) by sustaining a conformation that favors peptide exchange. RFLP analysis of HLA-DM genes from rheumatoid arthritis (RA) patients suggests that certain polymorphisms are genetic factors for RA susceptibility. HLA-B belongs to the HLA class I heavy chain paralogs. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. HLA-B and -C can form heterodimers consisting of a membrane anchored heavy chain and a light chain ( beta-2-Microglobulin). Polymorphisms yield hundreds of HLA-B and -C alleles. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| HLA-DRB3 | |
| Polyclonal | |
| Unconjugated | |
| HLA-DRB3 | |
| DR7; DRB1; DRw10; HLA class II histocompatibility antigen, DR beta 3 chain; HLA class II histocompatibility antigen, DRB1-7 beta chain; HLA DR3B; HLA-DR1B; HLA-DR3B; HLA-DRB; HLA-DRB3; human leucocyte antigen DRB3; major histocompatibility complex, class II, DR beta 3; MGC117330; MHC Class II antigen; MHC class II antigen DR beta 3 chain; MHC class II antigen DRB3; MHC class II HLA-DR beta 3 chain; SS1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3125 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P79483 | |
| HLA-DRB3 | |
| A synthesized peptide derived from human HLA-DRB3(Accession P79483), corresponding to amino acid residues A78-V128. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction