missing translation for 'onlineSavingsMsg'
Learn More

HLA DRB1 Antibody (SR1485), Novus Biologicals™

Rabbit Monoclonal Antibody

£257.00 - £341.00

Specifications

Antigen HLA DRB1
Clone SR1485
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18648634
View Documents
Novus Biologicals
NBP3-21924-100ul
100 μg
£341.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18644334
View Documents
Novus Biologicals
NBP3-21924-50ul
50 μg
£257.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HLA DRB1 Monoclonal antibody specifically detects HLA DRB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

HLA DRB1
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human
Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly
A synthesized peptide derived from human HLA DRB1 (Uniprot #: P01911)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1485
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
Adaptive Immunity, Asthma, Cell Biology, Diabetes Research, Immunology
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
3123
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.