missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DRA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17221-1-AP-150UL
This item is not returnable.
View return policy
Description
Expression of the invariant chain of the HLA-DR antigen (CD74) by B-lymphocytes begins in the pre-B stage but lost before the final differentiation to plasma cells. This protein is expressed in B-lymphocytes and many B-cell lymphomas.Specifications
| HLA-DRA | |
| Polyclonal | |
| Unconjugated | |
| HLA-DRA | |
| HLA DRA, HLA DRA1, HLA-DRA, MHC class II antigen DRA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3122 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01903 | |
| HLA-DRA | |
| HLA-DRA Fusion Protein Ag11044 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction