missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-DRA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111368
This item is not returnable.
View return policy
Description
Immunogen sequence: AIKEEHVIIQ AEFYLNPDQS GEFMFDFDGD EIFHVDMAKK Highest antigen sequence indentity to the following orthologs: Mouse - 41%, Rat - 83%.
HLA-DRA is one of the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha and a beta chain, both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The alpha chain is approximately 33-35 kDa and its gene contains 5 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain and the cytoplasmic tail. DRA does not have polymorphisms in the peptide binding part and acts as the sole alpha chain for DRB1, DRB3, DRB4 and DRB5. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| HLA-DRA | |
| Polyclonal | |
| Unconjugated | |
| HLA-DRA | |
| histocompatibility antigen HLA-DR alpha; HLA class II histocompatibility antigen, DR alpha chain; HLA-DRA; HLA-DRA1; major histocompatibility complex, class II, DR alpha; MHC cell surface glycoprotein; MHC class II antigen DRA; MLRW | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3122 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P01903 | |
| HLA-DRA | |
| Recombinant Protein corresonding to Human HLA-DRA. Recombinant protein control fragment (Product #RP-103147). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction