Learn More
HLA-DRA Mouse, anti-Human, Clone: 3D6E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68543-1-IG-150UL
This item is not returnable.
View return policy
Description
The HLA-DRA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HLA-DRA. This antibody recognizes human, pig antigen. The HLA-DRA antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| HLA-DRA | |
| 3D6E8 | |
| HLA DRA, HLA DRA1, HLA-DRA, MHC class II antigen DRA | |
| 150 μL | |
| 3122 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig | |
| 150 kDa | |
| IgG1 |