missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DRA Antibody (CL10964), Novus Biologicals™
Mouse Monoclonal Antibody
£239.00 - £494.00
Specifications
| Antigen | HLA DRA |
|---|---|
| Clone | CL10964 |
| Dilution | Western Blot 1 μg/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616447
|
Novus Biologicals
NBP3-07996-25ul |
25 μg |
£239.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620899
|
Novus Biologicals
NBP3-07996-100ul |
100 μg |
£494.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA DRA Monoclonal antibody specifically detects HLA DRA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| HLA DRA | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: EFYLNPDQSGEFMFDFDGDE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL10964 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology | |
| PBS, pH 7.2, containing 40% glycerol | |
| 3122 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title