missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DR/DP Antibody (HL-40), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-408AF350
This item is not returnable.
View return policy
Description
HLA DR/DP Monoclonal specifically detects HLA DR/DP in Human, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HLA DR/DP | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HLA-DRA | |
| Raji Burkitt′s lymphoma cell line | |
| Primary | |
| The antibody HL-40 reacts with common epitope on beta-chain of human HLA-DR and HLA-DP. DR and DP are the isotypes of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages). | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HL-40 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| Mouse | |
| 0.1 mL | |
| 3122 | |
| Human, Pig | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction