missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR Antibody (TAL 8.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2708-0.025mg
This item is not returnable.
View return policy
Description
HLA-DR Monoclonal antibody specifically detects HLA-DR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Radioimmune Assays, CyTOF
Specifications
| HLA-DR | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, CyTOF-ready | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| Cells from the B line WVB coated with mouse antiserum followed by WVB class II antigen. | |
| 0.025 mg | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
| 3122 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Radioimmune Assays (RIA) | |
| TAL 8.1 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction