missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR Antibody (HLA-DRA/6839R), Novus Biologicals™
Rabbit Monoclonal Antibody
£242.00 - £431.00
Specifications
| Antigen | HLA-DR |
|---|---|
| Clone | HLA-DRA/6839R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL, Immunohistochemistry-Frozen 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661594
|
Novus Biologicals
NBP3-20425-20ug |
20 μg |
£242.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653415
|
Novus Biologicals
NBP3-20425-100ug |
100 μg |
£431.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA-DR Monoclonal antibody specifically detects HLA-DR in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| HLA-DR | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
| 10mM PBS with 0.05% BSA | |
| 3122 | |
| IgG κ | |
| Protein A or G purified |
| HLA-DRA/6839R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL, Immunohistochemistry-Frozen 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| Recombinant fragment (around aa1-200) of human HLA-DR protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title