missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQW1 Antibody (Genox 3.53), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2702APCCY7
This item is not returnable.
View return policy
Description
HLA DQW1 Monoclonal specifically detects HLA DQW1 in Human samples. It is validated for Flow Cytometry.
Specifications
| HLA DQW1 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| HLA-DQB1 | |
| Bristol 8 glycoprotein from the Bristol 8 B lymphoblastoid cell line. | |
| 0.1 mL | |
| Primary | |
| Human Leukocyte Antigen DQW1 (DC1 determinant of human HLA-DR antigens) | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| Genox 3.53 | |
| Flow Cytometry | |
| CELIAC1, DC1, HLA class II histocompatibility antigen, DQ beta 1 chain, HLA-DQB, IDDM1, LB-E12, MB1, MHC class II antigen DQB1, MHC class II antigen HLA-DQ-beta-1, MHC class II DQ beta chain, MHC class II HLA-DQ beta glycoprotein, MT1 | |
| Mouse | |
| Protein A purified | |
| Asthma, Diabetes Research, Immunology | |
| 3119 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction