missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DQA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55072-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to HLA-DQA2.
HLA-DQA2 belongs to the HLA class II alpha chain family. The encoded protein forms a heterodimer with a class II beta chain. It is located in intracellular vesicles and plays a central role in the peptide loading of MHC class II molecules by helping to release the CLIP molecule from the peptide binding site. Class II molecules are expressed in antigen presenting cells (B lymphocytes, dendritic cells, macrophages) and are used to present antigenic peptides on the cell surface to be recognized by CD4 T-cells.Specifications
| HLA-DQA2 | |
| Polyclonal | |
| Unconjugated | |
| HLA-DQA2 | |
| DX alpha chain, HLA DQA1, HLA DQA2, HLA DXA, HLA-DQA2, MHC class II DQA2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3118 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01906 | |
| HLA-DQA2 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction