missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQ1/DQ3 Antibody (HL-37), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-405MFV500
This item is not returnable.
View return policy
Description
HLA DQ1/DQ3 Monoclonal antibody specifically detects HLA DQ1/DQ3 in Human, Porcine samples. It is validated for Flow Cytometry
Specifications
| HLA DQ1/DQ3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Pig | |
| Purified |
| Flow Cytometry | |
| HL-37 | |
| Flow Cytometry | |
| CELIAC1, HLA class II histocompatibility antigen, DQ beta 1 chain, HLA-DQB, IDDM1, MHC class II antigen DQB1, MHC class II antigen HLA-DQ-beta-1, MHC class II DQ beta chain, MHC class II HLA-DQ beta glycoprotein | |
| Burkitt's lymphoma cell line Raji | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Immunology | |
| 3119 | |
| Store at 4C in the dark. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction