missing translation for 'onlineSavingsMsg'
Learn More

HLA DQ Rabbit anti-Human, Porcine, Clone: HLA-DQA1/2866R, Novus Biologicals™

Product Code. 18371352 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371352 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371352 Supplier Novus Biologicals Supplier No. NBP30773220UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HLA DQ Monoclonal specifically detects HLA DQ in Human, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA DQ
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HLA-DQA1/2866R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Frozen 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Host Species Rabbit
Immunogen Recombinant full-length human HLA DQ protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3117
Target Species Human, Pig
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.