missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQ Mouse anti-Human, Porcine, Clone: rSPV-L3, Novus Biologicals™
Mouse Monoclonal Antibody
£415.00
Specifications
| Antigen | HLA DQ |
|---|---|
| Clone | rSPV-L3 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Frozen 1 to 2 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
Description
HLA DQ Monoclonal specifically detects HLA DQ in Human, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| HLA DQ | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Asthma, Immunology | |
| 10 mM PBS with 0.05% BSA | |
| 3117 | |
| IgG1 κ | |
| Protein A or G purified |
| rSPV-L3 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunohistochemistry-Frozen 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Pig | |
| CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
| T4-positive CTL clone HG-38 | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title