missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQ Mouse anti-Human, Clone: B-K27, Novus Biologicals™
Mouse Monoclonal Antibody
£324.00 - £610.00
Specifications
| Antigen | HLA DQ |
|---|---|
| Clone | B-K27 |
| Concentration | 1 mg/ml |
| Dilution | Flow Cytometry, Immunoprecipitation |
| Applications | Flow Cytometry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18336566
|
Novus Biologicals
NBP3-18113-200UL |
200 μg |
£324.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18324802
|
Bio-Techne
NBP3-18113-500UL |
500 μg |
£610.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA DQ Monoclonal antibody specifically detects HLA DQ in Human samples. It is validated for Flow Cytometry, ImmunoprecipitationSpecifications
| HLA DQ | |
| 1 mg/ml | |
| Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Asthma, Immunology | |
| PBS. Sterile-filtered through 0.22 um. | |
| 3117 | |
| IgG2a κ | |
| Affinity purified |
| B-K27 | |
| Flow Cytometry, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
| T4+ CTL clone HG-38. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title