missing translation for 'onlineSavingsMsg'
Learn More

HLA DQ/DR/DP, Rabbit anti-Human, Clone: HLA-Pan/2967R, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP2-79709-100UG

 View more versions of this product

Product Code. 18213086

  • £440.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HLA DQ/DR/DP Monoclonal specifically detects HLA DQ/DR/DP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HLA DQ/DR/DP
Monoclonal
0.2 mg/mL
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA 2-5 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
P04440, P01908, P01909, P01920, P01903, P01903
HLA-DQA1
Non-T, non-B human acute lymphoblastic leukemia REH6 cell line
100 μg
Adaptive Immunity, Asthma, Diabetes Research, Immunology
3117
Human
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
HLA-Pan/2967R
Unconjugated
10 mM PBS with 0.05% BSA
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Rabbit
Protein A or G purified
RUO
Primary
This antibody reacts with a common epitope of human major histocompatibility (MHC) class II antigens, HLA DQ/DR/DP.
Store at 4°C.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.