missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQ/DR/DP Mouse anti-Human, Clone: HLA-Pan/6663, Novus Biologicals™
Mouse Monoclonal Antibody
£216.00 - £416.00
Specifications
| Antigen | HLA DQ/DR/DP |
|---|---|
| Clone | HLA-Pan/6663 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18378405
|
Novus Biologicals
NBP3-13905-20UG |
20 μg |
£216.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18074145
|
Novus Biologicals
NBP3-13905-100UG |
100 μg |
£416.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA DQ/DR/DP Monoclonal antibody specifically detects HLA DQ/DR/DP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)Specifications
| HLA DQ/DR/DP | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Asthma, Diabetes Research, Immunology | |
| 10 mM PBS with 0.05% BSA | |
| CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
| Priess human B cell line | |
| Primary | |
| Store at 4°C. |
| HLA-Pan/6663 | |
| Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P04440, P01908, P01909, P01920, P01903 | |
| 3117 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title