missing translation for 'onlineSavingsMsg'
Learn More

HLA DQ/DR/DP Antibody (rHLA-Pan/8847), Novus Biologicals™

Mouse Monoclonal Antibody

£178.00 - £351.00

Specifications

Antigen HLA DQ/DR/DP
Clone rHLA-Pan/8847
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18623955
View Documents
Novus Biologicals
NBP3-23657-100ug
100 μg
£351.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18696844
View Documents
Novus Biologicals
NBP3-23657-20ug
20 μg
£178.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HLA DQ/DR/DP Monoclonal antibody specifically detects HLA DQ/DR/DP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

HLA DQ/DR/DP
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Adaptive Immunity, Asthma, Diabetes Research, Immunology
10 mM PBS with 0.05% BSA
3117
IgG1 κ
Protein A or G purified
rHLA-Pan/8847
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Priess human B cell line
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.