missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HLA-DPB1 Recombinant Rabbit Monoclonal Antibody (1X8F7)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558199

 View more versions of this product

Product Code. 30284161

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MMVLQVSAAP RTVALTALLM VLLTSVVQGR ATPENYLFQG RQECYAFNGT QRFLERYIYN REEFARFDSD VGEFRAVTEL GRPAAEYWNS QKDILEEKRA.

HLA-DPB belongs to the HLA class II beta chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DPA) and a beta chain (DPB), both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The beta chain is approximately 26-28 kDa and its gene contains 6 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and exon 5 encodes the cytoplasmic tail. Within the DP molecule both the alpha chain and the beta chain contain the polymorphisms specifying the peptide binding specificities, resulting in up to 4 different molecules. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HLA-DPB1
Recombinant Monoclonal
0.072 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Rabbit
Protein A
RUO
Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin)
1X8F7
Unconjugated
beta1 domain MHC class II HLA DPB; CD; CELIAC1; class II HLA beta chain; DC-1 alpha chain; DC-alpha; DPB1; DQ-A1; GSE; histocompatibility antigen HLA-DR alpha; HLA class II histocompatibility antigen, DP beta 1 chain; HLA class II histocompatibility antigen, DP(W4) beta chain; HLA class II histocompatibility antigen, DQ alpha 1 chain; HLA class II histocompatibility antigen, DQ(W3) alpha chain; HLA class II histocompatibility antigen, DR alpha chain; HLA DP14-beta chain; HLA-DCA; HLA-DP; HLA-DP histocompatibility type, beta-1 subunit; HLA-DP1B; HLA-DPB; HLA-DPB1; HLA-DQA; HLA-DQA1; HLA-DRA; HLA-DRA1; leucocyte antigen DQA1; leukocyte antigen alpha chain; major histocompatibility complex class II antigen beta chain; major histocompatibility complex, class II, DP beta 1; major histocompatibility complex, class II, DQ alpha 1; major histocompatibility complex, class II, DR alpha; MHC cell surface glycoprotein; MHC cla; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen DP beta 1 chain; MHC class II antigen DPB1; MHC class II antigen DPbeta1; MHC class II antigen DRA; MHC class II DQA1; MHC class II HLA-D alpha glycoprotein; MHC class II HLA-DP-beta-1; MHC class II HLA-DQ-alpha-1; MHC class II HLA-DRB1; MHC class II surface glycoprotein; MHC HLA DPB1; MHC HLA-DQ alpha; MLRW
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HLA-DPB1 (NP_002112.3).
20 μL
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.