Learn More
Invitrogen™ HLA-DPA1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5101635
Product Code. 16628114
  • £444.00 / 100µL
Description
Antibody detects endogenous levels of total HLA-DPA1.
HLA-DPA1 belongs to the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DPA) and a beta (DPB) chain, both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The alpha chain is approximately 33-35 kDa and its gene contains 5 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and the cytoplasmic tail. Within the DP molecule both the alpha chain and the beta chain contain the polymorphisms specifying the peptide binding specificities, resulting in up to 4 different molecules. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HLA-DPA1
Polyclonal
Unconjugated
HLA-DPA1
DP(W3); DP(W4); HLA class II histocompatibility antigen, DP alpha 1 chain; HLADP; HLA-DP1A; HLA-DPA1; HLASB; HLA-SB alpha chain; major histocompatibility complex, class II, DP alpha 1; MHC class II antigen; MHC class II DP3-alpha; MHC class II DPA1; MHC class II HLA-DPA1 antigen; PLT1; Primed lymphocyte test-1
Rabbit
Affinity chromatography
RUO
3113
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P20036
HLA-DPA1
A synthesized peptide derived from human HLA-DPA1(Accession P20036), corresponding to amino acid residues F43-N93.
100 μL
Primary
Human, Mouse, Rat