missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DP Mouse anti-Human, PE, Clone: B7/21 (also known as B7/21.49, B7/21 (IgG1)), BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566825
This item is not returnable.
View return policy
Description
HLA-DP Monoclonal antibody specifically detects HLA-DP in Human samples. It is validated for Flow Cytometry.
Specifications
| HLA-DP | |
| Monoclonal | |
| PE | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| B7/21 (also known as B7/21.49, B7/21 (IgG1)) | |
| Hu HLA-DP PE B7/21 100Tst | |
| Human NALM6 Leukemia Cells | |
| RUO | |
| HLA-DP | |
| Refrigerate (2 to 8°C) | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction