missing translation for 'onlineSavingsMsg'
Learn More

HLA-DP/-DQ/-DR Mouse, Clone: CR3/43, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 3115-MSM3-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15970168

  • £395.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HLA-DP/-DQ/-DR
Cell surface
CR3/43
Unconjugated
NP_002112
Mouse
36kDa (? chain) and 27kDa (? chain)
Protein A/G Purified
RUO
3115
IgG1
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
HLA-DP ;HLA-DQ ;HLA-DR
Cells from human tonsil
Raji cells. Tonsil or lymph node
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers