missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DOB Antibody (DOB 10E6/5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£159.00 - £302.00
Specifications
| Antigen | HLA DOB |
|---|---|
| Clone | DOB 10E6/5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635516
|
Novus Biologicals
NBP2-50420-0.025ml |
0.025 mL |
£159.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656108
|
Novus Biologicals
NBP2-50420 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA DOB Monoclonal antibody specifically detects HLA DOB in Human samples. It is validated for Western Blot, ELISASpecifications
| HLA DOB | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Asthma, Immunology | |
| PBS | |
| 3112 | |
| IgG1 κ | |
| Protein A purified |
| DOB 10E6/5 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DOB, FLJ57033, HLA class II histocompatibility antigen, DO beta chain, major histocompatibility complex, class II, DO beta, MHC class II antigen DOB | |
| C-terminal peptide MHC class II | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title