missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HLA DOA Polyclonal specifically detects HLA DOA in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | HLA DOA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | HLA class II histocompatibility antigen, DO alpha chain, HLA-D0-alpha, HLA-DNAHLADZ, HLA-DZA, lymphocyte antigen, major histocompatibility complex, class II, DN alpha, major histocompatibility complex, class II, DO alpha, MHC class II antigen DOA, MHC DN-alpha, MHC DZ alpha |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 26-135 of human HLA DOA (NP_002110.1). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?