missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DMB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | HLA-DMB |
|---|---|
| Concentration | 0.38 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16831305
|
Proteintech
21704-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16821305
|
Proteintech
21704-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA-DMB (major histocompatibility complex, class II, DM beta), also known as D6S221E, RING7, HLA-DM histocompatibility type, beta chain, HLADMB or RING7, is a protein that in humans is encoded by the HLA-DMB gene. The HLA-DMB gene is mapped on 6p21.32. HLA-DMB belongs to the HLA class II beta chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DMA) and a beta (DMB) chain, both anchored in the membrane. It is located in intracellular vesicles. DM plays a central role in the peptide loading of MHC class II molecules by helping to release the CLIP (class II-associated invariant chain peptide) molecule from the peptide binding site. Class II molecules are expressed in antigen presenting cells. The beta chain is approximately 26-28 kDa and its gene contains 6 exons. HLA-DMA and -DMB appear to encode subunits of a functional heterodimer that is critical in the pathway of class II antigen presentation.Specifications
| HLA-DMB | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P28068 | |
| 3109 | |
| HLA-DMB Fusion Protein Ag16320 | |
| Primary | |
| -20°C | |
| HLA-DMB |
| 0.38 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| D6S221E, DMB, HLA DMB, HLA-DMB, MHC class II antigen DMB, RING7 | |
| HLA-DMB | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title