missing translation for 'onlineSavingsMsg'
Learn More

HLA DMB Rabbit anti-Human, Clone: 3C2X4, Novus Biologicals™

Product Code. 18360745 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360745 100 μg 100µL
18374692 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18360745 Supplier Novus Biologicals Supplier No. NBP315325100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HLA DMB Monoclonal antibody specifically detects HLA DMB in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA DMB
Applications Western Blot
Classification Monoclonal
Clone 3C2X4
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias class II histocompatibility antigen, M beta chain, D6S221E, DMB, major histocompatibility complex, class II, DM beta, MHC class II antigen DMB, MHC class II antigen HLA-DM beta chain, MHC class II HLA-DMB, Really interesting new gene 7 protein, RING7HLA class II histocompatibility antigen, DM beta chain
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HLA DMB (P28068).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Asthma, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3109
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.