missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HLA-DMA Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA564969

Product Code. 15977744

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LEFGKPNTLVC FVSNLFPPML TVNWQHHSVP VEGFGPTFVS AVDGLSFQAF SYLNFTPEPS DIFSCIVTHE IDRYTAIAYW VPRNALPSDL LENV Highest antigen sequence identity to the following orthologs - mouse 84%, rat 85%.

HLA-DMA belongs to the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DMA) and a beta chain (DMB), both anchored in the membrane. It is located in intracellular vesicles. DM plays a central role in the peptide loading of MHC class II molecules by helping to release the CLIP molecule from the peptide binding site. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The alpha chain is approximately 33-35 kDa and its gene contains 5 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and the cytoplasmic tail. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HLA-DMA
Polyclonal
Unconjugated
HLA-DMA
class II histocompatibility antigen, M alpha chain; D6S222E; DAAP-27A1.5; DMA; HLA class II histocompatibility antigen, DM alpha chain; HLADM; HLA-DMA; major histocompatibility complex, class II, DM alpha; MHC class II antigen DMA; really interesting new gene 6 protein; RING6
Rabbit
Antigen affinity chromatography
RUO
3108
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P28067
HLA-DMA
Recombinant Human HLA-DMA. Recombinant protein control fragment (Product #RP-105741).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.