missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DMA Antibody (6B12NB), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44302JF549
This item is not returnable.
View return policy
Description
HLA DMA Monoclonal antibody specifically detects HLA DMA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Cell Surface)
Specifications
| HLA DMA | |
| Monoclonal | |
| Janelia Fluor 549 | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| Partial recombinant human HLA DMA protein (between amino acids 10-300) [Entrez NP_006111] | |
| 0.1 mL | |
| Asthma, Diabetes Research, Immunology, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroendocrinology | |
| 3108 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry (Cell Surface) | |
| 6B12NB | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction