missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA Class I Antibody (MEM-123), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-505PE
This item is not returnable.
View return policy
Description
HLA Class I Monoclonal specifically detects HLA Class I in Human, Bovine, Primate samples. It is validated for Flow Cytometry.
Specifications
| HLA Class I | |
| Monoclonal | |
| PE | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody (clone MEM-123) reacts with all human classical MHC Class I molecules (major histocompatibility complex) in native cell-surface forms as well as with human HLA-G cDNA transfected cells. MHC Class I molecules (MHC Class Ia) are expressed on the surface of all human cell types. This antibody completely blocks binding of classical W6/32 to surface-expressed HLA-G, but does not cross-blocks This antibody MEM-G/9. | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| MEM-123 | |
| Flow Cytometry | |
| HLA-A | |
| COS-7 African green monkey kidney cells | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 3105 | |
| Human, Bovine, Primate | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction