missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA C Antibody (DT-9), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50419R
This item is not returnable.
View return policy
Description
HLA C Monoclonal antibody specifically detects HLA C in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| HLA C | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HLA-C | |
| Purified tamarin MHC class I molecules (using anti-class I antibody BB7.7) | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3107 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| DT-9 | |
| Flow Cytometry | |
| C alpha chain, D6S204, FLJ27082, HLA class I histocompatibility antigen, Cw-1 alpha chain, HLAC, HLA-Cw, HLA-JY3, HLC-C, human leukocyte antigen-C alpha chain, leucocyte antigen C, leucocyte antigen Cw, leukocyte antigen-C alpha chain, major histocompatibility antigen HLA-C, major histocompatibility complex, class I, C, MHC class I antigen Cw*1, MHC class I antigen Cw*12, MHC class I antigen Cw*14, MHC class I antigen Cw*15, MHC class I antigen Cw*16, MHC class I antigen Cw*17, MHC class I antigen Cw*2, MHC class I antigen Cw*3, MHC class I antigen Cw*4, MHC class I antigen Cw*5, MHC class I antigen Cw*6, MHC class I antigen Cw*7, MHC class I antigen Cw*8, MHC class I antigen heavy chain HLA-C, PSORS1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction