missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA C Antibody (DT-9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£174.00 - £335.00
Specifications
| Antigen | HLA C |
|---|---|
| Clone | DT-9 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
| Applications | Flow Cytometry, Immunoprecipitation, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629706
|
Novus Biologicals
NBP2-50419-0.025ml |
0.025 mL |
£174.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670978
|
Novus Biologicals
NBP2-50419 |
0.1 mL |
£335.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA C Monoclonal antibody specifically detects HLA C in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOFSpecifications
| HLA C | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunoprecipitation, CyTOF | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immunology | |
| PBS | |
| 3107 | |
| IgG2b | |
| Protein A purified |
| DT-9 | |
| Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C alpha chain, D6S204, FLJ27082, HLA class I histocompatibility antigen, Cw-1 alpha chain, HLAC, HLA-Cw, HLA-JY3, HLC-C, human leukocyte antigen-C alpha chain, leucocyte antigen C, leucocyte antigen Cw, leukocyte antigen-C alpha chain, major histocompatibility antigen HLA-C, major histocompatibility complex, class I, C, MHC class I antigen Cw*1, MHC class I antigen Cw*12, MHC class I antigen Cw*14, MHC class I antigen Cw*15, MHC class I antigen Cw*16, MHC class I antigen Cw*17, MHC class I antigen Cw*2, MHC class I antigen Cw*3, MHC class I antigen Cw*4, MHC class I antigen Cw*5, MHC class I antigen Cw*6, MHC class I antigen Cw*7, MHC class I antigen Cw*8, MHC class I antigen heavy chain HLA-C, PSORS1 | |
| Purified tamarin MHC class I molecules (using anti-class I antibody BB7.7) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title